Thermo Scientific™

Pierce™ Gaussia Luciferase Flash Assay Kit

Catalog number: 16158
Have Questions?
Thermo Scientific™

Pierce™ Gaussia Luciferase Flash Assay Kit

Catalog number: 16158
Have Questions?
Catalog Number
16158
Unit Size
100 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1000 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
16158100 reactions
-
161591000 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Thermo Scientific Pierce Gaussia Luciferase Flash Assay Kit provides researchers with the most sensitive assay for transcriptional activity of regulatory elements in mammalian cell culture media and whole cell lysate.

Features of the Gaussia Luciferase Flash Assay Kit:

Sensitive—1000 times greater sensitivity allows utilization of smaller numbers of cells
Cost effective—highest sensitivity assays result in decreased reagent consumption
Secreted luciferase—allows real-time assays and kinetic studies without destroying cells
Time-saving—assays using secreted luciferases require minimal sample handling
Automation-friendly—amenable to high throughput screens
Convenient—contains a universal cell lysis buffer and optimized flash assay reagent
Safe—allows one to perform non-radioactive assays

This Flash Assay Kit contains reagents for measuring the activity of Gaussia luciferase in mammalian cell culture media and lysates. When used with Thermo Scientific Gaussia Luc Vectors, the kit provides an extremely sensitive bioluminescent reporter assay system for secreted or intracellular detection of promoter or pathway activity. Gaussia luciferase is highly secreted into the cell culture media, allowing for live cell monitoring of reporter activity. The signal produced by Gaussia is considerably greater than signal from either firefly or Renilla luciferases assayed under similar conditions.

Includes:
Cell lysis buffer, reaction buffer and substrate

Requires:
Gaussia luciferase and luminometer or other instrument capable of monitoring luminescence, such as Thermo Scientific Luminoskan Ascent and Varioskan Flash Microplate Readers.

Applications:
• Promoter studies for analyzing cis- regulatory elements and trans-acting factors
• Drug screening
• siRNA and miRNA screening
• Multiplexed assays to study off-target effects
• Secretory pathway/protein localization reporter assays
• Signal transduction pathway analysis
• RNA splicing studies

Gaussia luciferase is a 20kDa protein from the marine copepod, Gaussia princeps. The bioluminescent enzyme is highly secreted into the cell culture media, allowing for live cell monitoring of reporter activity. Light output generated by the luciferase reaction can be correlated to the amount of Gaussia luciferase protein produced and used to determine the activity of the promoter driving Gaussia expression. The signal produced by Gaussia luciferase shows strong flash kinetics and is considerably greater than flash signals from either firefly or Renilla luciferases assayed under similar conditions. The assay kit is optimized for use with our Gaussia Luc Vectors but is fully compatible with other Gaussia luciferases and derivatives that use coelenterazine as a substrate. (For glow assays, use our Gaussia-Dura Luc Vectors.)

More Product Data
Highly sensitive multiplex luciferase reporter assays
Luciferase assays in hard-to-transfect Jurkat cells
Monitoring neuronal differentiation using multiplexed luciferase reporters
Activation of the antioxidant response pathway by pesticide chemicals
Versatile luciferases: microplate luminometers and flash luciferase assays
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Type
Assay Kit
Target
Luciferase, Gaussia Luciferase
Sufficient For
100 Luciferase Assays
Quantity
100 Reaction Kit
Assay
Reporter Enzyme, Luciferase Reporter Assay
Compatible Cells
Mammalian Cells
Detection Method
Bioluminescent
For Use With (Equipment)
Luminometer (Microplate)
Format
384-well plate, 96-well plate
Label Type
Enzyme Labeled
Substrate Properties
Chemical Substrate
Technique
Enhanced Chemiluminescence
Substrate Type
Luciferase Substrate
Product Line
Pierce™
Substrate
Coelenterazine

Contents & Storage

Sufficient For: 100 luciferase assays in microplate wells
• Gaussia Flash Assay Buffer, 5 mL (store at 4°C)
• 100X Coelenterazine, 50 μL (store at -80°C)
• Luciferase Cell Lysis Buffer (2X), 6 mL (store at room temperature)

Kit may be stored at -80°C, or individually at indicated temperatures.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text