Thermo Scientific™

Pierce™ Renilla Luciferase Flash Assay Kit

Catalog number: 16164
Have Questions?
Thermo Scientific™

Pierce™ Renilla Luciferase Flash Assay Kit

Catalog number: 16164
Have Questions?
Catalog Number
16164
Unit Size
100 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1000 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
16164100 reactions
-
161651000 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Thermo Scientific Pierce Renilla Luciferase Flash Assay Kit provides researchers with a highly sensitive intracellular assay for transcriptional activity of regulatory elements in mammalian whole cell lysate.

Features of the Renilla Luciferase Flash Assay Kit:

Sensitive—greater sensitivity allows utilization of smaller numbers of cells
Cost effective—highest sensitivity assays result in decreased reagent consumption
Time-saving—assays require minimal sample handling
Automation-friendly—amenable to high throughput screens
Convenient—contains a universal cell lysis buffer and optimized flash assay reagent
Safe—allows one to perform non-radioactive assays

This Flash Assay Kit contains reagents for measuring the activity of Renilla luciferase in mammalian cell lysates. When used with Thermo Scientific Green Renilla Luc Plasmids, the kit provides an extremely sensitive bioluminescent reporter assay system for intracellular detection of promoter or pathway activity. The signal produced by Green Renilla Luc is considerably greater than signal from either native firefly or Renilla luciferases assayed under similar conditions. Light output captured using a luminometer can be correlated with the amount of Renilla luciferase protein produced and used to determine the activity of the promoter driving Renilla expression.

Includes:
Cell lysis buffer, reaction buffer and substrate

Requires:
Renilla luciferase and luminometer or other instrument capable of monitoring luminescence, such as Thermo Scientific Luminoskan Ascent and Varioskan Flash Microplate Readers.

Applications:
• Promoter studies for analyzing cis- regulatory elements and trans-acting factors
• Drug screening
• siRNA and miRNA screening
• Multiplexed assays to study off-target effects
• Protein localization reporter assays
• Signal transduction pathway analysis
• RNA splicing studies

The Green Renilla luciferase is a 36kDa protein produced by a derivative of the wild type Renilla luciferase gene from the sea pansy, Renilla reniformis. Compared to the wild type luciferase, Green Renilla is more stable in serum and has an the emission spectrum that is shifted toward the green region. The protein provides extremely bright flash signal that decays rapidly. The Pierce Renilla Flash Assay Kit reagent optimizes the this signal for use in flash-type luciferase reporter assays. The kit also is compatible for flash assays with other Renilla luciferases and derivatives that use coelenterazine as a substrate.

More Product Data
Luciferase assays in hard-to-transfect Jurkat cells
Versatile luciferases: microplate luminometers and flash luciferase assays

Related Products
Pierce™ Renilla-Firefly Luciferase Dual Assay Kit
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Type
Assay Kit
Target
Luciferase, Green Renilla Luciferase
Quantity
100 Reaction Kit
Assay
Reporter Enzyme, Luciferase Reporter Assay
Compatible Cells
Mammalian Cells
Detection Method
Bioluminescent
For Use With (Equipment)
Luminometer (Microplate)
Format
384-well plate, 96-well plate
Label Type
Enzyme Labeled
Substrate Properties
Chemical Substrate
Technique
Enhanced Chemiluminescence
Substrate Type
Luciferase Substrate
Product Line
Pierce™
Substrate
Coelenterazine

Contents & Storage

• Renilla Flash Assay Buffer, 5 mL, store at 4°C
• Coelenterazine (100X), 50 μL, store at -80°C
• 2X Cell Lysis Buffer, 6 mL, store at room temperature

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text