Thermo Scientific™

Pierce™ Cypridina Luciferase Flash Assay Kit

Catalog number: 16168
Have Questions?
Thermo Scientific™

Pierce™ Cypridina Luciferase Flash Assay Kit

Catalog number: 16168
Have Questions?
Catalog Number
16168
Unit Size
100 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1000 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
16168100 reactions
-
161691000 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Thermo Scientific Pierce Cypridina Luciferase Flash Assay Kit provides researchers with a highly sensitive assay for transcriptional activity of regulatory elements in mammalian cell culture media and whole cell lysate.

Features of the Cypridina Luciferase Flash Assay Kit:

Sensitive—high sensitivity allows utilization of smaller numbers of cells
Cost effective—sensitive assays result in decreased reagent consumption
Secreted luciferase—allows real-time assays and kinetic studies without destroying cells
Ideal for co-transfection—uses a different substrate than Renilla, Gaussia, or firefly luciferases
Compatible—assay reagents compatible with other Cypridina or Vargula luciferases
Time-saving—assays using secreted luciferases require minimal sample handling
Automation-friendly—amenable to high throughput screens
Convenient—contains a universal cell lysis buffer and optimized flash assay reagent
Safe—allows one to perform non-radioactive assays

This Flash Assay Kit contains reagents for measuring the activity of Cypridina luciferase in mammalian cell culture media and lysates. When used with Thermo Scientific Cypridina Luc Vectors, the kit provides an extremely sensitive bioluminescent reporter assay system for secreted or intracellular detection of promoter or pathway activity. Cypridina luciferase is highly secreted into the cell culture media, allowing for live cell monitoring of reporter activity. The signal produced by Cypridina is considerably greater than signal from either firefly or Renilla luciferases assayed under similar conditions.

Includes:
Cell lysis buffer, reaction buffer and substrate

Requires:
Cypridina luciferase and luminometer or other instrument capable of monitoring luminescence, such as Thermo Scientific Luminoskan Ascent and Varioskan Flash Microplate Readers.

Applications:
• Promoter studies for analyzing cis- regulatory elements and trans-acting factors
• Drug screening
• siRNA and miRNA screening
• Multiplexed assays to study off-target effects
• Secretory pathway/protein localization reporter assays
• Signal transduction pathway analysis
• RNA splicing studies

The Cypridina luciferase is a 61kDa protein from the marine ostracod, Cypridina noctiluca. Cypridina luciferase is highly secreted into the cell culture media, allowing for live cell monitoring of reporter activity. Light output captured using a luminometer can be correlated with the amount of Cypridina luciferase protein produced and used to determine the activity of the promoter driving Cypridina expression. The Cypridina gene in Thermo Scientific Cypridina Luc Vectors is modified for expression in mammalian cells.

The assay kit is optimized for use with our Cypridina Luc Vectors but is fully compatible with any luciferases that use Vargulin as a substrate. Because Cypridina uses a different substrate than Renilla, Gaussia, or firefly luciferases, co-transfection and assays with up to three reporters are possible. The signal produced by Cypridina shows strong flash kinetics and is considerably greater than flash signals from either firefly or Renilla luciferases assayed under similar conditions.

More Product Data
Highly sensitive multiplex luciferase reporter assays
Luciferase assays in hard-to-transfect Jurkat cells
Monitoring neuronal differentiation using multiplexed luciferase reporters
Activation of the antioxidant response pathway by pesticide chemicals
Versatile luciferases: microplate luminometers and flash luciferase assays
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Type
Assay Kit
Target
Luciferase, Cypridina Luciferase
Quantity
100 Reaction Kit
Assay
Reporter Enzyme, Luciferase Reporter Assay
Compatible Cells
Mammalian Cells
Detection Method
Bioluminescent
For Use With (Equipment)
Luminometer (Microplate)
Format
384-well plate, 96-well plate
Label Type
Enzyme Labeled
Substrate Properties
Chemical Substrate
Technique
Enhanced Chemiluminescence
Substrate Type
Luciferase Substrate
Product Line
Pierce™
Substrate
Vargulin (Cypridina Luciferin)

Contents & Storage

• Cypridina Flash Assay Buffer, 5 mL, store at 4°C
• Vargulin (1X), 5 μL, store at -20°C
• 2X Cell Lysis Buffer, 6 mL, store at room temperature

Upon receipt store kit at -20°C or store individual components as indicated above.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text