Thermo Scientific Chemicals

4,4'-Dimethoxybenzophenone, 97%, Thermo Scientific Chemicals

Catalog number: 161741000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

4,4'-Dimethoxybenzophenone, 97%, Thermo Scientific Chemicals

Catalog number: 161741000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 161741000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
90-96-0
IUPAC Name
bis(4-methoxyphenyl)methanone
Molecular Formula
C15H14O3
InChI Key
RFVHVYKVRGKLNK-UHFFFAOYSA-N
SMILES
COC1=CC=C(C=C1)C(=O)C1=CC=C(OC)C=C1
Appearance (Color)
Yellow to light beige
Appearance (Form)
Powder
Infrared spectrum
Conforms
HPLC
>=96.0 %
Melting point
141°C to 146°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text