Thermo Scientific™

Pierce™ Gaussia-Firefly Luciferase Dual Assay Kit

Catalog number: 16182
Have Questions?
Thermo Scientific™

Pierce™ Gaussia-Firefly Luciferase Dual Assay Kit

Catalog number: 16182
Have Questions?
Catalog Number
16182
Unit Size
1000 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
100 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
161821000 reactions
-
16181100 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Thermo Scientific Pierce Gaussia-Firefly Luciferase Dual Assay Kit provides the necessary reagents to simultaneously detect intracellular Gaussia and Red Firefly luciferase activity in mammalian whole cell lysates.

Features of the Gaussia-Firefly Luciferase Dual Assay Kit:

Simultaneous—concurrent, filter-based, wavelength-separated detection of two luciferase activities
Sensitive—measure bright blue Gaussia and red firefly luciferase activities in the same sample
Fast—no quenching step required, unlike in traditional sequential dual assays
Multiplex—capable of quantitating two cellular activities in the same sample(s)

The Pierce Gaussia-Firefly Luciferase Dual Assay Kit is a highly sensitive assay that allows for the simultaneous detection of Gaussia and Red Firefly luciferase activity. Gaussia luciferase acts as an experimental reporter with constitutively active red firefly as a normalization control. As shown in the animation videos below, this reporter-and-control combination enables simultaneous monitoring of experimental reporter and control luciferase activities in a single-read assay without the need for two-step addition of substrate reagents or quenching. The assay working solution contains substrates for both luciferases, and the reactions occur simultaneously with flash-type kinetics. The resulting luminescent signals are spectrally resolvable using filters. In a single sample, researchers can assay transcriptional activity of regulatory elements, signal transduction pathways, and effects of activators or inhibitors.

Includes:
Cell lysis buffer, reaction buffer, and substrates coelenterazine and D-luciferin

Requires:
Gaussia and Red Firefly luciferase reporters; Filter sets in the ranges of 425 to 525nm for Gaussia Luc and 615 to 675nm for Red Firefly Luc. Luminometer or other instrument capable of monitoring luminescence, such as Thermo Scientific Luminoskan Ascent and Varioskan Flash Microplate Readers; Injector required for assessing more than 24 wells at a time.

Applications:
• Study two regulatory elements at the same time
• Monitor two signaling pathways simultaneously
• Enable studying more than one target per screen (e.g. off-target effects)

Application Videos:
Gaussia luciferase produces brighter bioluminescent signal than firefly and native Renilla luciferases. The bioluminescent signal (λmax= 470nm) produced by Gaussia luciferase results from the oxidation of coelenterazine. This reaction does not require adenosine triphosphate (ATP) or other cofactors. Red firefly luciferase is a mutant form of the Japanese firefly luciferase from Luciola cruciata. This luciferase produces a red-shifted emission spectrum (λmax= 613nm) that results from the oxidation of D-luciferin in the presence of ATP. Red firefly and Gaussia luciferases are used in the dual assay because the light output is spectrally resolvable.

More Product Data
Highly sensitive multiplex luciferase reporter assays
Simultaneous dual-emission detection of luciferase reporter assays
Activation of the antioxidant response pathway by pesticide chemicals
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Type
Assay Kit
Target
Luciferase, Firefly Luciferase, Gaussia Luciferase
Quantity
1000 Reaction Kit
Assay
Reporter Enzyme, Luciferase Reporter Assay
Compatible Cells
Mammalian Cells
Detection Method
Bioluminescent
For Use With (Equipment)
Luminometer (Microplate)
Format
384-well plate, 96-well plate
Label Type
Enzyme Labeled
Substrate Properties
Chemical Substrate
Technique
Enhanced Chemiluminescence
Substrate Type
Luciferase Substrate
Product Line
Pierce™
Substrate
Coelenterazine, D-Luciferin

Contents & Storage

• Gaussia-Firefly Dual Assay Buffer, 50 mL, store at 4°C
• Coelenterazine (100X), 0.5 mL, store at -80°C
• D-Luciferin, Lyophilized, 30 mg, store at 4°C
• 2X Cell Lysis Buffer, 60 mL, store at room temperature

Upon receipt store kit at -80°C or store individual components as indicated above.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text