Thermo Scientific Chemicals

N,N-Dichlorourethane 98%, Thermo Scientific Chemicals

Catalog number: 161820050
5 g, Glass Bottle, Each
Thermo Scientific Chemicals

N,N-Dichlorourethane 98%, Thermo Scientific Chemicals

Catalog number: 161820050
5 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 161820050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
13698-16-3
IUPAC Name
ethyl N,N-dichlorocarbamate
Molecular Formula
C3H5Cl2NO2
InChI Key
PCGGNOFZZFFFLU-UHFFFAOYSA-N
SMILES
CCOC(=O)N(Cl)Cl
GC
>=97.5 %
Refractive index
1.4590 to 1.4610 (20°C, 589 nm)
Appearance (Color)
Clear yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text