2,4,6-Collidine, 99%, Thermo Scientific Chemicals
2,4,6-Collidine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,4,6-Collidine, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
1 L
2.5 L
Catalog number 161951000
Price (USD)/ Each
86.65
Online exclusive
96.10 
Save 9.45 (10%)
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
86.65
Online exclusive
96.10 
Save 9.45 (10%)
Add to cart
2,4,6-Collidine, 99%, Thermo Scientific Chemicals
Catalog number161951000
Price (USD)/ Each
86.65
Online exclusive
96.10 
Save 9.45 (10%)
-
Add to cart
Chemical Identifiers
CAS108-75-8
IUPAC Name2,4,6-trimethylpyridine
Molecular FormulaC8H11N
InChI KeyBWZVCCNYKMEVEX-UHFFFAOYSA-N
SMILESCC1=CC(C)=NC(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4970 to 1.4990 (20°C, 589 nm)
AppearanceMay darken on storage
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only