Thermo Scientific Chemicals

2,4,6-Collidine, 99%, Thermo Scientific Chemicals

Catalog number: 161955000
500 mL, Each
Thermo Scientific Chemicals

2,4,6-Collidine, 99%, Thermo Scientific Chemicals

Catalog number: 161955000
500 mL, Each
Quantity
Catalog number: 161955000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
108-75-8
IUPAC Name
2,4,6-trimethylpyridine
Molecular Formula
C8H11N
InChI Key
BWZVCCNYKMEVEX-UHFFFAOYSA-N
SMILES
CC1=CC(C)=NC(C)=C1
Appearance (Color)
Clear colorless to yellow
Infrared spectrum
Conforms
GC
>=98.5 %
Refractive index
1.4970 to 1.4990 (20°C, 589 nm)
Appearance
May darken on storage

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text