Phenylglyoxal monohydrate, 97%, Thermo Scientific Chemicals
Phenylglyoxal monohydrate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Phenylglyoxal monohydrate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 161970050
Price (USD)/ Each
92.70
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
92.70
Add to cart
Phenylglyoxal monohydrate, 97%, Thermo Scientific Chemicals
Catalog number161970050
Price (USD)/ Each
92.70
-
Add to cart
Chemical Identifiers
IUPAC Name2,2-dihydroxy-1-phenylethan-1-one
Molecular FormulaC8H8O3
InChI KeyNBIBDIKAOBCFJN-UHFFFAOYSA-N
SMILESOC(O)C(=O)C1=CC=CC=C1
Molecular Weight (g/mol)152.15
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow to pink
Titration excess NaOH>=96.0 %
Appearance (Form)Powder
Infrared spectrumConforms
Melting point71°C to 76°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only