Thermo Scientific Chemicals

3-Chloropropylamine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 162012500
250 g, Plastic bottle, Each
Thermo Scientific Chemicals

3-Chloropropylamine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 162012500
250 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 162012500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
6276-54-6
IUPAC Name
hydrogen 3-chloropropan-1-amine chloride
Molecular Formula
C3H9Cl2N
InChI Key
IHPRVZKJZGXTBQ-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NCCCCl
Titration Argentometric
>=98 % (Ionised Chloride)
Appearance (Form)
Crystalline powder and/or chunks
Infrared spectrum
Conforms
Titration Argentometric
>=97.5 % (Total Chloride)
Water
=<0.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text