Thermo Scientific Chemicals

Phenyl benzoate, 99%, Thermo Scientific Chemicals

Catalog number: 162065000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Phenyl benzoate, 99%, Thermo Scientific Chemicals

Catalog number: 162065000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 162065000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
93-99-2
IUPAC Name
phenyl benzoate
Molecular Formula
C13H10O2
InChI Key
FCJSHPDYVMKCHI-UHFFFAOYSA-N
SMILES
O=C(OC1=CC=CC=C1)C1=CC=CC=C1
Appearance (Form)
Crystalline powder
Melting point
68°C to 70°C
Appearance (Color)
White
Infrared spectrum
Conforms
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text