N,N-Dimethylformamide diethyl acetal, 95%, Thermo Scientific Chemicals
N,N-Dimethylformamide diethyl acetal, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N,N-Dimethylformamide diethyl acetal, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 162080100
Price (USD)/ Each
-
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS1188-33-6
IUPAC Name(diethoxymethyl)dimethylamine
Molecular FormulaC7H17NO2
InChI KeyBWKAYBPLDRWMCJ-UHFFFAOYSA-N
SMILESCCOC(OCC)N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.3990 to 1.4010 (20°C, 589 nm)
Appearance (Form)Liquid
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
GC>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
RUO – Research Use Only