Thermo Scientific Chemicals

Difluoroacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: 162120500
50 mL, Glass bottle, Each
Thermo Scientific Chemicals

Difluoroacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: 162120500
50 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 162120500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
381-73-7
IUPAC Name
2,2-difluoroacetic acid
Molecular Formula
C2H2F2O2
InChI Key
PBWZKZYHONABLN-UHFFFAOYSA-N
SMILES
OC(=O)C(F)F
Appearance (Color)
Clear colorless to light brown
Infrared spectrum
Conforms
Titration with NaOH
>=97.5 %
Refractive index
1.3420 to 1.3450 (20°C, 589 nm)
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text