Appearance (Form)Crystalline powder or crystals and/or chunks
Appearance (Color)White to brown
Infrared spectrumConforms
Titration RCOCl>=96.0 %
Melting point57°C to 61°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.