2,2'-Dithiodipyridine, 98%, Thermo Scientific Chemicals
2,2'-Dithiodipyridine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,2'-Dithiodipyridine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 162230050
Price (USD)/ Each
95.65
Online exclusive
106.00 
Save 10.35 (10%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
95.65
Online exclusive
106.00 
Save 10.35 (10%)
Add to cart
2,2'-Dithiodipyridine, 98%, Thermo Scientific Chemicals
Catalog number162230050
Price (USD)/ Each
95.65
Online exclusive
106.00 
Save 10.35 (10%)
-
Add to cart
Chemical Identifiers
CAS2127-03-9
IUPAC Name2-(pyridin-2-yldisulfanyl)pyridine
Molecular FormulaC10H8N2S2
InChI KeyHAXFWIACAGNFHA-UHFFFAOYSA-N
SMILESS(SC1=CC=CC=N1)C1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to beige
Melting point55°C to 60°C
Infrared spectrumConforms
GC>=97.5 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only