Thermo Scientific Chemicals

2,2'-Dithiodipyridine, 98%, Thermo Scientific Chemicals

Catalog number: 162231000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

2,2'-Dithiodipyridine, 98%, Thermo Scientific Chemicals

Catalog number: 162231000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 162231000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2127-03-9
IUPAC Name
2-(pyridin-2-yldisulfanyl)pyridine
Molecular Formula
C10H8N2S2
InChI Key
HAXFWIACAGNFHA-UHFFFAOYSA-N
SMILES
S(SC1=CC=CC=N1)C1=CC=CC=N1
Appearance (Color)
White to yellow to beige
Melting point
55°C to 60°C
Infrared spectrum
Conforms
GC
>=97.5 %
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text