Diglycolic acid, 98%, Thermo Scientific Chemicals
Diglycolic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diglycolic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 162265000
Price (USD)/ Each
-
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS110-99-6
IUPAC Name2-(carboxymethoxy)acetic acid
Molecular FormulaC4H6O5
InChI KeyQEVGZEDELICMKH-UHFFFAOYSA-N
SMILESOC(=O)COCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with NaOH>=97.5 %
Appearance (Form)Crystalline powder
Melting point141°C to 145°C
Appearance (Color)White to light beige
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only