Homovanillic acid, 98%, Thermo Scientific Chemicals
Homovanillic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Homovanillic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 162530050
Price (USD)/ Each
228.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
269.00 
Save 40.35 (15%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
228.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
269.00 
Save 40.35 (15%)
Add to cart
Homovanillic acid, 98%, Thermo Scientific Chemicals
Catalog number162530050
Price (USD)/ Each
228.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
269.00 
Save 40.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name2-(4-hydroxy-3-methoxyphenyl)acetic acid
Molecular FormulaC9H10O4
InChI KeyQRMZSPFSDQBLIX-UHFFFAOYSA-N
SMILESCOC1=CC(CC(O)=O)=CC=C1O
Molecular Weight (g/mol)182.18
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
HPLC>=97.5 %
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point141°C to 145°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only