Iminostilbene, 97%, Thermo Scientific Chemicals
Iminostilbene, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Iminostilbene, 97%, Thermo Scientific Chemicals

Have Questions?
Quantity:
100 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 162541000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS256-96-2
IUPAC Name2-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,9,11,13-heptaene
Molecular FormulaC14H11N
InChI KeyLCGTWRLJTMHIQZ-UHFFFAOYSA-N
SMILESN1C2=CC=CC=C2C=CC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange
GC>=96.0 %
Melting point196°C to 202°C
Infrared spectrumConforms
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only