Thermo Scientific Chemicals

3-Hydroxy-4-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: 162571000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

3-Hydroxy-4-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: 162571000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 162571000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
621-59-0
IUPAC Name
3-hydroxy-4-methoxybenzaldehyde
Molecular Formula
C8H8O3
InChI Key
JVTZFYYHCGSXJV-UHFFFAOYSA-N
SMILES
COC1=CC=C(C=O)C=C1O
Melting point
112°C to 116°C
Appearance (Form)
Powder
Infrared spectrum
Conforms
GC
>=97.5 %
Appearance (Color)
Off-white to grey to light brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text