Propionamide, 97%
Propionamide, 97%
Propionamide, 97%
Thermo Scientific Chemicals

Propionamide, 97%

CAS: 79-05-0 | C3H7NO | 73.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Catalog number 162670250
Price (USD)
63.65
Online Exclusive
70.00
Save 6.35 (9%)
Each
Estimated availability date  Pending  
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
63.65
Online Exclusive
70.00
Save 6.35 (9%)
Each
Add to cart
Chemical Identifiers
CAS79-05-0
IUPAC Namepropanamide
Molecular FormulaC3H7NO
InChI KeyQLNJFJADRCOGBJ-UHFFFAOYSA-N
SMILESCCC(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point77°C to 83°C
Appearance (Color)Colorless to white to light yellow
Infrared spectrumConforms
GC>=96.0 %
Appearance (Form)Powder or crystals or flakes and/or chunks
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link