Thermo Scientific Chemicals

Propionamide, 97%, Thermo Scientific Chemicals

Catalog number: 162671000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Propionamide, 97%, Thermo Scientific Chemicals

Catalog number: 162671000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 162671000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
79-05-0
IUPAC Name
propanamide
Molecular Formula
C3H7NO
InChI Key
QLNJFJADRCOGBJ-UHFFFAOYSA-N
SMILES
CCC(N)=O
Appearance (Color)
Colorless to white to light yellow
Infrared spectrum
Conforms
GC
>=96.0 %
Appearance (Form)
Powder or crystals or flakes and/or chunks
Melting point
77°C to 83°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text