Thermo Scientific Chemicals

δ-Valerolactone, 99%, Thermo Scientific Chemicals

Catalog number: 162710500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

δ-Valerolactone, 99%, Thermo Scientific Chemicals

Catalog number: 162710500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 162710500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
542-28-9
IUPAC Name
oxan-2-one
Molecular Formula
C5H8O2
InChI Key
OZJPLYNZGCXSJM-UHFFFAOYSA-N
SMILES
O=C1CCCCO1
Infrared spectrum
Conforms
Appearance (Form)
Liquid
Additional info
polymerized product
Appearance (Color)
Clear colorless to light yellow
Additional info
remark: may contain variable amounts of

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text