δ-Valerolactone, 99%, Thermo Scientific Chemicals
δ-Valerolactone, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

δ-Valerolactone, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1 kg
Packaging:
Glass bottle
Catalog number 162712500
Price (USD)/ Each
-
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Nameoxan-2-one
Molecular FormulaC5H8O2
InChI KeyOZJPLYNZGCXSJM-UHFFFAOYSA-N
SMILESO=C1CCCCO1
Molecular Weight (g/mol)100.12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Additional inforemark: may contain variable amounts of
Appearance (Form)Liquid
Additional infopolymerized product
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only