Thermo Scientific Chemicals

4-Hydroxybenzaldehyde, 99%, Thermo Scientific Chemicals

Catalog number: 162770010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

4-Hydroxybenzaldehyde, 99%, Thermo Scientific Chemicals

Catalog number: 162770010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 162770010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
123-08-0
IUPAC Name
4-hydroxybenzaldehyde
Molecular Formula
C7H6O2
InChI Key
RGHHSNMVTDWUBI-UHFFFAOYSA-N
SMILES
OC1=CC=C(C=O)C=C1
GC
>=98.5 %
Melting point
114°C to 118°C
Infrared spectrum
Conforms
Appearance (Color)
Yellow to light brown
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text