4-Hydroxybenzaldehyde, 99%, Thermo Scientific Chemicals
4-Hydroxybenzaldehyde, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Hydroxybenzaldehyde, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
50 g
250 g
1 kg
Packaging:
Plastic bottle
Catalog number 162770025
Price (USD)/ Each
-
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS123-08-0
IUPAC Name4-hydroxybenzaldehyde
Molecular FormulaC7H6O2
InChI KeyRGHHSNMVTDWUBI-UHFFFAOYSA-N
SMILESOC1=CC=C(C=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point114°C to 118°C
Infrared spectrumConforms
GC>=98.5 %
Appearance (Color)Yellow to light brown
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only