4-Hydroxybenzaldehyde, 99%
4-Hydroxybenzaldehyde, 99%
4-Hydroxybenzaldehyde, 99%
4-Hydroxybenzaldehyde, 99%
Thermo Scientific Chemicals

4-Hydroxybenzaldehyde, 99%

CAS: 123-08-0 | C7H6O2 | 122.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Catalog number 162770500
Price (USD)
31.65
Special offer
Online exclusive
Ends: 26-Sep-2025
39.40
Save 7.75 (20%)
Each
In stock
Add to cart
Quantity:
50 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
31.65
Special offer
Online exclusive
Ends: 26-Sep-2025
39.40
Save 7.75 (20%)
Each
Add to cart
Chemical Identifiers
CAS123-08-0
IUPAC Name4-hydroxybenzaldehyde
Molecular FormulaC7H6O2
InChI KeyRGHHSNMVTDWUBI-UHFFFAOYSA-N
SMILESOC1=CC=C(C=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=98.5 %
Melting point114°C to 118°C
Appearance (Color)Yellow to light brown
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection