Thermo Scientific Chemicals

chlorodimethylsilane, 96%, Thermo Scientific Chemicals

Catalog number: 162840100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

chlorodimethylsilane, 96%, Thermo Scientific Chemicals

Catalog number: 162840100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 162840100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1066-35-9
IUPAC Name
chlorodimethylsilyl
Molecular Formula
C2H6ClSi
InChI Key
QABCGOSYZHCPGN-UHFFFAOYSA-N
SMILES
C[Si](C)Cl
Infrared spectrum
Conforms
Refractive index
1.3815 to 1.3855 (20°C, 589 nm)
Appearance
May develop some turbidity or precipitate
GC
>=96.0 %
Appearance (Color)
Clear to slightly hazy colorless to light yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text