chlorodimethylsilane, 96%, Thermo Scientific Chemicals
chlorodimethylsilane, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

chlorodimethylsilane, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
10 g
50 g
Packaging:
Glass bottle
Catalog number 162842500
Price (USD)/ Each
178.65
Online exclusive
210.00 
Save 31.35 (15%)
-
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
178.65
Online exclusive
210.00 
Save 31.35 (15%)
Add to cart
chlorodimethylsilane, 96%, Thermo Scientific Chemicals
Catalog number162842500
Price (USD)/ Each
178.65
Online exclusive
210.00 
Save 31.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namechlorodimethylsilyl
Molecular FormulaC2H6ClSi
InChI KeyQABCGOSYZHCPGN-UHFFFAOYSA-N
SMILESC[Si](C)Cl
Molecular Weight (g/mol)93.61
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.3815 to 1.3855 (20°C, 589 nm)
AppearanceMay develop some turbidity or precipitate
GC>=96.0 %
Appearance (Color)Clear to slightly hazy colorless to light yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only