4-Amino-m-cresol, 99+%, Thermo Scientific Chemicals
4-Amino-m-cresol, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Amino-m-cresol, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Packaging:
Glass bottle
Catalog number 163202500
Price (USD)/ Each
-
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2835-99-6
IUPAC Name4-amino-3-methylphenol
Molecular FormulaC7H9NO
InChI KeyQGNGOGOOPUYKMC-UHFFFAOYSA-N
SMILESCC1=CC(O)=CC=C1N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to yellow to beige to brown
Infrared spectrumConforms
Appearance (Form)Powder or crystals and/or chunks
Melting point176°C to 180°C
GC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only