2-Amino-p-cresol, 97%, Thermo Scientific Chemicals
2-Amino-p-cresol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Amino-p-cresol, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Packaging:
Glass bottle
Catalog number 163210500
Price (USD)/ Each
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS95-84-1
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Infrared spectrumConforms
Appearance (Form)Crystalline powder or flakes
Melting point134°C to 139°C
Appearance (Color)Beige to grey-brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only