96 Well Round (U) Bottom Plate, TC Surface, Pack of 1
Save 51% on plasticware essentials and stock your lab to culture with confidence. View promotion details >
96 Well Round (U) Bottom Plate, TC Surface, Pack of 1
Thermo Scientific™

96 Well Round (U) Bottom Plate, TC Surface, Pack of 1

Ideal in a wide variety of applications, including cloning experiments and tissue culture-based analytical systems
Have Questions?
Catalog NumberQuantity
163320Case of 50
Catalog number 163320
Price (USD)
383.65
Online Exclusive
410.00
Save 26.35 (6%)
Case of 50
In stock
Add to cart
Quantity:
Case of 50
Price (USD)
383.65
Online Exclusive
410.00
Save 26.35 (6%)
Case of 50
Add to cart

Thermo Scientific™ Nunc MicroWell Plates with Nunclon Delta Surface are ideal in a wide variety of applications, including cloning experiments and tissue culture-based analytical systems. Thermo Scientific Nunc Cell Culture products offer solutions for cell growth and analysis and have been used by researchers worldwide since 1953. With a wide range of surface areas and treatments, we have products for all your basic or advanced cell-based research.

  • Tight-fitting lids reduce evaporation
  • Stackable
  • Radiation sterilized
  • Condensation rings control evaporation and retain condensate
  • Keyed lids prevent accidental reversals
  • Dimensions: 86 × 128mm

Ordering Information:

Individually wrapped.

WARNING: Cancer – www.P65Warnings.ca.gov
Specifications
Certifications/ComplianceAll microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
CertifiedNunclon Delta certified for monolayer formation, cloning efficiency, non-cytotoxic, non-pyrogenic, and sterility
LidWith Lid
No. per Case50
No. per Pack1
Packaging1/Pk., 50/Cs.
Product TypeMicroplate
QuantityCase of 50
Array8 x 12
ColorClear
MaterialPolystyrene
No. of Wells96
Product LineNunc
SterilitySterile
Surface TreatmentNunclon Delta
Volume (Metric) Well300 μL
Well ShapeRound Bottom (U-well)
Unit SizeCase of 50

Frequently asked questions (FAQs)

What is the difference between the Nunclon Delta surface and a tissue culture (TC) surface from a competitor?

Our Nunclon Delta surface for adherent cells is a standard tissue culture (TC) surface modification that makes the polystyrene surface more hydrophilic. This facilitates maximum adhesion for a broad range of cell types.

The Nunclon Delta surface is Thermo Scientific Nunclon Delta-certified, which is a Thermo Fisher proprietary certification that has a quality standard to help ensure consistency. Every lot of Thermo Scientific Nunclon Delta-certified cell cultureware is tested with Gibco media for:
- Less than 10% growth variation in L929, Hel299 and primary chick embryo cells
- Less than 15% variation colony formation of V79-4 cells

More information and all applicable products can be found in the "Product selection guide for every step of your cell culture workflow" brochure .

What reagents are used to coat your Nunclon Delta surface products?

The Nunclon Delta surface is a surface modification that enhances cell attachment and growth for adherent cell lines and is not coated with any chemical reagent.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link