Thermo Scientific Chemicals

1,3-Dihydroxynaphthalene, 99+%, Thermo Scientific Chemicals

Catalog number: 163340250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

1,3-Dihydroxynaphthalene, 99+%, Thermo Scientific Chemicals

Catalog number: 163340250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 163340250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
132-86-5
IUPAC Name
naphthalene-1,3-diol
Molecular Formula
C10H8O2
InChI Key
XOOMNEFVDUTJPP-UHFFFAOYSA-N
SMILES
OC1=CC(O)=C2C=CC=CC2=C1
Appearance (Color)
Pink-orange to brown
Appearance (Form)
Crystalline powder
Melting point
123°C to 126°C
Titration with NaOH
>=99.0 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text