Thermo Scientific Chemicals

4,6-Dichloropyrimidine, 97%, Thermo Scientific Chemicals

Catalog number: 163371000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

4,6-Dichloropyrimidine, 97%, Thermo Scientific Chemicals

Catalog number: 163371000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 163371000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1193-21-1
IUPAC Name
4,6-dichloropyrimidine
Molecular Formula
C4H2Cl2N2
InChI Key
XJPZKYIHCLDXST-UHFFFAOYSA-N
SMILES
ClC1=CC(Cl)=NC=N1
Melting point
64°C to 70°C
Infrared spectrum
Conforms
Appearance (Color)
White to yellow to orange
Appearance (Form)
Powder or crystals or needles
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text