2,3-Dimethyl-1,3-butadiene, 98%, stabilized with BHT, Thermo Scientific Chemicals
2,3-Dimethyl-1,3-butadiene, 98%, stabilized with BHT, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,3-Dimethyl-1,3-butadiene, 98%, stabilized with BHT, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 163430050
Price (USD)/ Each
43.70
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
43.70
Add to cart
2,3-Dimethyl-1,3-butadiene, 98%, stabilized with BHT, Thermo Scientific Chemicals
Catalog number163430050
Price (USD)/ Each
43.70
-
Add to cart
Chemical Identifiers
CAS513-81-5
SpecificationsSpecification SheetSpecification Sheet
Stabilizer100 ppm BHT
Appearance (Form)Liquid
Refractive index1.4370 to 1.4400 (20°C, 589 nm)
Appearance (Color)Clear colorless
GC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only