Thermo Scientific Chemicals

Brucine dihydrate, 99%, Thermo Scientific Chemicals

Catalog number: 163500500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

Brucine dihydrate, 99%, Thermo Scientific Chemicals

Catalog number: 163500500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 163500500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
145428-94-0
Titration with HClO4
>=98.5 % (On dry substance)
Appearance (Form)
Powder or crystals
Melting point
174°C to 179°C
Loss on drying
6.0 % to 9.5 % (105°C, 3 hrs)
Appearance (Color)
White to light beige

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text