1-Bromodecane, 98%, Thermo Scientific Chemicals
1-Bromodecane, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Bromodecane, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
Packaging:
Glass bottle
Catalog number 163605000
Price (USD)/ Each
49.65
Online exclusive
58.80 
Save 9.15 (16%)
-
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
49.65
Online exclusive
58.80 
Save 9.15 (16%)
Add to cart
1-Bromodecane, 98%, Thermo Scientific Chemicals
Catalog number163605000
Price (USD)/ Each
49.65
Online exclusive
58.80 
Save 9.15 (16%)
-
Add to cart
Chemical Identifiers
IUPAC Name1-bromodecane
Molecular FormulaC10H21Br
InChI KeyMYMSJFSOOQERIO-UHFFFAOYSA-N
SMILESCCCCCCCCCCBr
Molecular Weight (g/mol)221.18
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC97.5 %
Appearance (Color)Clear colorless to light yellow
Refractive index1.4550 to 1.4570 (20°C, 589 nm)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only