2-Bromopropionyl chloride, 98%, Thermo Scientific Chemicals
2-Bromopropionyl chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Bromopropionyl chloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
10 mL
50 mL
Packaging:
Glass bottle
Catalog number 163622500
Price (USD)/ Each
-
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7148-74-5
IUPAC Name2-bromopropanoyl chloride
Molecular FormulaC3H4BrClO
InChI KeyOZGMODDEIHYPRY-UHFFFAOYNA-N
SMILESCC(Br)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.4770 to 1.4810 (20°C, 589 nm)
Titration Argentometric>=97.5 % (ex Cl)
Infrared spectrumConforms
Appearance (Color)Clear yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only