Thermo Scientific Chemicals

Adenine sulfate, 98%, synthetic

Catalog number: 163631000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Adenine sulfate, 98%, synthetic

Catalog number: 163631000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 163631000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
321-30-2
IUPAC Name
bis(7H-purin-6-amine); sulfuric acid
Molecular Formula
C10H12N10O4S
InChI Key
LQXHSCOPYJCOMD-UHFFFAOYSA-N
SMILES
OS(O)(=O)=O.NC1=C2NC=NC2=NC=N1.NC1=C2NC=NC2=NC=N1
Appearance (Form)
Powder
UV spectrophotometry
molar absorptivity 13100 +/- 2%, on dry basis
Titration with NaOH
>=97.5 %
Loss on drying
=<2 % (1 g, 105°C, 5 hrs) (vacuum)
Sulfated ash
=<0.1 % (800°C)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text