Quinine sulfate dihydrate, 99+%
Quinine sulfate dihydrate, 99+%
Thermo Scientific Chemicals

Quinine sulfate dihydrate, 99+%

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number 163710500
Price (USD)/ Each
302.65
Online exclusive
336.00 
Save 33.35 (10%)
-
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
302.65
Online exclusive
336.00 
Save 33.35 (10%)
Add to cart
Quinine sulfate dihydrate, 99+%
Catalog number163710500
Price (USD)/ Each
302.65
Online exclusive
336.00 
Save 33.35 (10%)
-
Add to cart
Chemical Identifiers
CAS6119-70-6
IUPAC Namebis((R)-[(2S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](6-methoxyquinolin-4-yl)methanol) sulfuric acid dihydrate
Molecular FormulaC40H54N4O10S
InChI KeyZHNFLHYOFXQIOW-OIGVVMIYNA-N
SMILESO.O.OS(O)(=O)=O.[H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C.[H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Residue after ignition=<0.1 %
Specific optical rotation-235° to -245° (20°C, 589 nm) (c=2, 0.1N HCl)
Specific optical rotation(on anhydrous basis)
Appearance (Form)Powder or crystals or needles
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only