Thermo Scientific Chemicals

β-Alanine, 99%

Catalog number: 163790100
10 kg, Plastic Drum, Each
Thermo Scientific Chemicals

β-Alanine, 99%

Catalog number: 163790100
10 kg, Plastic Drum, Each
Quantity
Packaging
Catalog number: 163790100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
107-95-9
IUPAC Name
3-aminopropanoic acid
Molecular Formula
C3H7NO2
InChI Key
UCMIRNVEIXFBKS-UHFFFAOYSA-N
SMILES
NCCC(O)=O
Appearance (Color)
White
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Titration with HClO4
98.5 to 101.5 % (on dry substance)
Loss on drying
=<0.5 % (105°C, 3 h)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text