Thermo Scientific Chemicals

2,2-Bis(hydroxymethyl)-2,2',2″-nitrilotriethanol, 98%, Thermo Scientific Chemicals

Catalog number: 163825000
500 g, Each
Thermo Scientific Chemicals

2,2-Bis(hydroxymethyl)-2,2',2″-nitrilotriethanol, 98%, Thermo Scientific Chemicals

Catalog number: 163825000
500 g, Each
Quantity
Catalog number: 163825000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6976-37-0
IUPAC Name
2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
Molecular Formula
C8H19NO5
InChI Key
OWMVSZAMULFTJU-UHFFFAOYSA-N
SMILES
OCCN(CCO)C(CO)(CO)CO
Appearance (Form)
Powder
Titration with HClO4
>=97.5 %
Appearance (Color)
White to off-white
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text