Silicone oil, for melting point and boiling point apparatuses
Silicone oil, for melting point and boiling point apparatuses
Silicone oil, for melting point and boiling point apparatuses
Silicone oil, for melting point and boiling point apparatuses
Thermo Scientific Chemicals

Silicone oil, for melting point and boiling point apparatuses

CAS: 63148-62-9 | (C2H6OSi)n | NaN g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
2.5 kg
Packaging:
Glass bottle
Catalog number 163850025
Price (USD)
272.65
Online Exclusive
303.00
Save 30.35 (10%)
Each
In stock
Add to cart
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
272.65
Online Exclusive
303.00
Save 30.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS63148-62-9
IUPAC NamePolydimethylsiloxane
Molecular Formula(C2H6OSi)n
SMILESC[Si](C)(-*)O-*
MDL NumberMFCD00132673
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive index1.4030 to 1.4060 (20°C, 589 nm)
Appearance (Form)Oily liquid
Viscosity475 to 525 mm²/s (at 25°C)
Volatile matter=<1.5 % (5 g, 230°C, 2 h)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection