Thermo Scientific Chemicals

Silicone oil, for melting point and boiling point apparatuses, Thermo Scientific Chemicals

Catalog number: 163855000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Silicone oil, for melting point and boiling point apparatuses, Thermo Scientific Chemicals

Catalog number: 163855000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 163855000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
63148-62-9
IUPAC Name
Polydimethylsiloxane
Molecular Formula
(C2H6OSi)n
SMILES
C[Si](C)(-*)O-*
MDL Number
MFCD00132673
Appearance (Color)
Clear colorless
Infrared spectrum
Conforms
Refractive index
1.4030 to 1.4060 (20°C, 589 nm)
Appearance (Form)
Oily liquid
Viscosity
475 to 525 mm²/s (at 25°C)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text