Thermo Scientific Chemicals

7-Amino-1,3-naphthalenedisulfonic acid, Tech., Thermo Scientific Chemicals

Catalog number: 163960025
2.5 kg, Plastic Bottle, Each
Thermo Scientific Chemicals

7-Amino-1,3-naphthalenedisulfonic acid, Tech., Thermo Scientific Chemicals

Catalog number: 163960025
2.5 kg, Plastic Bottle, Each
Quantity
Packaging
Catalog number: 163960025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
86-65-7
IUPAC Name
7-aminonaphthalene-1,3-disulfonic acid
Molecular Formula
C10H9NO6S2
InChI Key
CMOLPZZVECHXKN-UHFFFAOYSA-N
SMILES
NC1=CC=C2C=C(C=C(C2=C1)S(O)(=O)=O)S(O)(=O)=O
Appearance (Color)
Off-white to green-beige to light brown
Infrared spectrum
Conforms
Appearance (Form)
Powder or needles
Titration with NaOH
>=60 % (As is)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text