Informational:Informational:This product ships via ground from US northeast.
Thermo Scientific Chemicals

Adenosine, 99+%

Catalog number: 164045000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Adenosine, 99+%

Catalog number: 164045000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 164045000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
58-61-7
IUPAC Name
(2R,3R,4S,5R)-2-(6-amino-9H-purin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
Molecular Formula
C10H13N5O4
InChI Key
OIRDTQYFTABQOQ-KQYNXXCUSA-N
SMILES
NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC=N1
Appearance (Color)
White
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Melting point
233°C to 238°C
UV spectrophotometry
98.0 to 102.0 % (lambda max. 259 nm, pH 7)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text