2-Hydroxy-5-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals
2-Hydroxy-5-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Hydroxy-5-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
5 g
10 g
Packaging:
Glass bottle
Catalog number 164160500
Price (USD)/ Each
427.00
-
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
427.00
Add to cart
2-Hydroxy-5-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals
Catalog number164160500
Price (USD)/ Each
427.00
-
Add to cart
Chemical Identifiers
CAS672-13-9
IUPAC Name2-hydroxy-5-methoxybenzaldehyde
Molecular FormulaC8H8O3
InChI KeyFZHSPPYCNDYIKD-UHFFFAOYSA-N
SMILESCOC1=CC=C(O)C(C=O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.5770 to 1.5790 (20°C, 589 nm)
Appearance (Color)Clear yellow
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only