Tetrahydrofuran, 99.6%, ACS reagent, stabilized with BHT, Thermo Scientific Chemicals
Tetrahydrofuran, 99.6%, ACS reagent, stabilized with BHT, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetrahydrofuran, 99.6%, ACS reagent, stabilized with BHT, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 L
1 L
2.5 L
25 L
Packaging:
View all
Plastic Bottle
Catalog number 164240050
Price (USD)/ Each
207.65
Online exclusive
231.00 
Save 23.35 (10%)
-
Add to cart
Quantity:
5 L
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)/ Each
207.65
Online exclusive
231.00 
Save 23.35 (10%)
Add to cart
Tetrahydrofuran, 99.6%, ACS reagent, stabilized with BHT, Thermo Scientific Chemicals
Catalog number164240050
Price (USD)/ Each
207.65
Online exclusive
231.00 
Save 23.35 (10%)
-
Add to cart
Chemical Identifiers
CAS109-99-9
IUPAC Nameoxolane
Molecular FormulaC4H8O
InChI KeyWYURNTSHIVDZCO-UHFFFAOYSA-N
SMILESC1CCOC1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.5 %
Residue after ignition=<0.002 %
Copper (Cu)=<0.05 ppm
Iron (Fe)=<0.5 ppm
Color scale=<10 APHA
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only