Acrylic acid, 98%, extra pure, stabilized
Acrylic acid, 98%, extra pure, stabilized
Acrylic acid, 98%, extra pure, stabilized
Acrylic acid, 98%, extra pure, stabilized
Thermo Scientific Chemicals

Acrylic acid, 98%, extra pure, stabilized

CAS: 79-10-7 | C3H4O2 | 72.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
View all
Glass bottle
Catalog number 164250010
Price (USD)
-
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS79-10-7
IUPAC Nameprop-2-enoic acid
Molecular FormulaC3H4O2
InChI KeyNIXOWILDQLNWCW-UHFFFAOYSA-N
SMILESOC(=O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<20 APHA
Refractive index1.4190 to 1.4230 (20°C, 589 nm)
Infrared spectrumConforms
Water=<0.1 % (K.F.)
Stabilizer180 to 220 ppm MEHQ
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0469941Certificate of AnalysisMay 21, 2025164250010, 164250100, 164252500, 164250025
A0469399Certificate of AnalysisMay 17, 2025164250010, 164250100, 164252500, 164250025
A0464678Certificate of AnalysisJan 31, 2025164250010, 164250100, 164252500, 164250025
A0464678Certificate of AnalysisJul 26, 2024164250010, 164250100, 164252500, 164250025
A0459748Certificate of AnalysisFeb 21, 2024164250010, 164250100, 164252500, 164250025
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link