Thermo Scientific Chemicals

Acrylic acid, 98%, extra pure, stabilized, Thermo Scientific Chemicals

Catalog number: 164252500
250 mL, Glass bottle, Each
Thermo Scientific Chemicals

Acrylic acid, 98%, extra pure, stabilized, Thermo Scientific Chemicals

Catalog number: 164252500
250 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 164252500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
79-10-7
IUPAC Name
prop-2-enoic acid
Molecular Formula
C3H4O2
InChI Key
NIXOWILDQLNWCW-UHFFFAOYSA-N
SMILES
OC(=O)C=C
Appearance (Form)
Clear liquid
GC
>=97.5 %
Dimer
=<2.5 % diacrylic acid
Color scale
=<20 APHA
Refractive index
1.4190 to 1.4230 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text